Tisk…

7-Chloroquinoline

Dodavatel: Apollo Scientific
Varování

OR14909-1G OR14909-5G OR14909-25G
APOSOR14909-1GEA 622 CZK
APOSOR14909-1G APOSOR14909-5G APOSOR14909-25G
7-Chloroquinoline
7-Chloroquinoline
Vzorec: C₉H₆ClN
Storage Temperature: Ambient
MDL Number: MFCD00956370
CAS číslo: 612-61-3
EINECS: 612-108-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR