(+)-Di-p-toluoyl-D-tartaric acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid
OR14900-100G
OR14900-250G
OR14900-500G
APOSOR14900-100GEA
622
CZK
APOSOR14900-100G
APOSOR14900-250G
APOSOR14900-500G
(+)-Di-p-toluoyl-D-tartaric acid 99%
(+)-Di-p-toluoyl-D-tartaric acid
Optical resolving agent.
Vzorec:
C₂₀H₁₈O₈ MW: 386.36 g/mol Bod tání: 169…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008552 CAS číslo: 32634-68-7 EINECS: 251-132-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...