Tisk…

2-Chlorothiazole-5-carbaldehyde 98%

Dodavatel: Apollo Scientific
Varování

OR14891-1G OR14891-250MG OR14891-5G
APOSOR14891-1GEA 705 CZK
APOSOR14891-1G APOSOR14891-250MG APOSOR14891-5G
2-Chlorothiazole-5-carbaldehyde 98%
2-Chlorothiazole-5-carbaldehyde
Vzorec: C₄H₂ClNOS
Storage Temperature: Refrigerator
MDL Number: MFCD01568717
CAS číslo: 95453-58-0
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR