Tisk…

2-Methylfuran-3-boronic acid pinacol ester 90%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane

OR14882-1G OR14882-250MG OR14882-5G
APOSOR14882-1GEA 5720 CZK
APOSOR14882-1G APOSOR14882-250MG APOSOR14882-5G
2-Methylfuran-3-boronic acid pinacol ester 90%
2-Methylfuran-3-boronic acid pinacol ester
Vzorec: C₁₁H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD09879905
CAS číslo: 864776-02-3
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR