2-Chloro-1,3-thiazole-5-carboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR14876-1G
OR14876-5G
APOSOR14876-1GEA
953
CZK
APOSOR14876-1G
APOSOR14876-5G
2-Chloro-1,3-thiazole-5-carboxylic acid 97%
2-Chloro-1,3-thiazole-5-carboxylic acid
Vzorec:
C₄H₂ClNO₂S Bod tání: 165…167 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02187331 CAS číslo: 101012-12-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...