5-Amino-3-methyl-1,2,4-thiadiazole
Dodavatel: Apollo Scientific
|
Varování
|
OR14846-1G
OR14846-5G
OR14846-10G
APOSOR14846-1GEA
622
CZK
APOSOR14846-1G
APOSOR14846-5G
APOSOR14846-10G
5-Amino-3-methyl-1,2,4-thiadiazole
5-Amino-3-methyl-1,2,4-thiadiazole
Vzorec:
C₃H₅N₃S Storage Temperature: Ambient |
MDL Number:
MFCD01114751 CAS číslo: 17467-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...