tert-Butyl-3-[4-(methoxycarbonyl)phenyl]-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate
Dodavatel: Apollo Scientific
Synonyms:
2,2-Dioxo-3-[4-(methoxycarbonyl)phenyl]diazathiane, N1-BOC protected
OR14832-10G
OR14832-1G
OR14832-5G
APOSOR14832-10GEA
25460
CZK
APOSOR14832-10G
APOSOR14832-1G
APOSOR14832-5G
tert-Butyl-3-[4-(methoxycarbonyl)phenyl]-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate
tert-Butyl-3-[4-(methoxycarbonyl)phenyl]-2,2-dioxo-2lambda~6~-diazathiane-1-carboxylate
Vzorec:
C₁₃H₁₈N₂O₆S MW: 330,36 g/mol Bod tání: 135…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD09865024 CAS číslo: 1017782-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...