Tisk…

1-[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl]-1-ethanol

Dodavatel: Apollo Scientific

Synonyms: 1-(4-Bromobenzyl)-4-(1-hydroxyethyl)-5-methyl-1H-1,2,3-triazole

OR14809-1G OR14809-5G
APOSOR14809-1GEA 9560 CZK
APOSOR14809-1G APOSOR14809-5G
1-[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl]-1-ethanol
1-[1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazol-4-yl]-1-ethanol
Vzorec: C₁₂H₁₄BrN₃O
MW: 296,17 g/mol
Bod tání: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD09865012
CAS číslo: 1017782-60-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR