1H-Pyrazole-1-carboximidamide hydrochloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR14754-100G
OR14754-25G
APOSOR14754-100GEA
2110
CZK
APOSOR14754-100G
APOSOR14754-25G
1H-Pyrazole-1-carboximidamide hydrochloride
1H-Pyrazole-1-carboximidamide hydrochloride
Vzorec:
C₄H₇ClN₄ Bod tání: 167…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210087 CAS číslo: 4023-02-3 EINECS: 429-520-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...