Tisk…

1-(3-Chloropropyl)-4-methylpiperazine dihydrochloride

Dodavatel: Apollo Scientific
Varování

OR14748-10G OR14748-1G OR14748-5G
APOSOR14748-10GEA 8070 CZK
APOSOR14748-10G APOSOR14748-1G APOSOR14748-5G
1-(3-Chloropropyl)-4-methylpiperazine dihydrochloride
1-(3-Chloropropyl)-4-methylpiperazine dihydrochloride
Vzorec: C₈H₁₉Cl₃N₂
Storage Temperature: Ambient
CAS číslo: 2031-23-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR