Tisk…

Triacetyl resveratrol 98+%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (E)-5-(4-Acetoxystyryl)-1,3-phenylene diacetate, 3,5,4′-Tri-O-acetylresveratrol, Acetyl-trans-resveratrol, 5-[(1E)-2-[4-(acetyloxy)phenyl]ethenyl]-1,3-Benzenediol-1,3-diacetate

OR14747-5G OR14747-10G
APOSOR14747-5GEA 622 CZK
APOSOR14747-5G APOSOR14747-10G
Triacetyl resveratrol 98+%
Triacetyl resveratrol
Vzorec: C₂₀H₁₈O₆
MW: 354,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01546481
CAS číslo: 42206-94-0
UN: 3077
ADR: 9+EHS,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR