L-Allylglycine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(S)-(-)-alpha-Allylglycine, H-Ally-Gly-OH, H-Gly(Ally)-OH, 2-Allyl-L-glycine, (S)-(-)-2-Amino-4-pentenoic acid, (2S)-2-Aminopent-4-enoic acid, L-2-Amino-4-pentenoic acid L-Allylglycine L-alpha-Allylglycine, (S)-2-Allylglycine
OR14736-5G
APOSOR14736-5GEA
1080
CZK
APOSOR14736-5G
L-Allylglycine
L-Allylglycine
Vzorec:
C₅H₉NO₂ MW: 115,132 g/mol Bod tání: 283 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00002627 CAS číslo: 16338-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...