Tisk…

3-(2-Thienyl)-L-alanine 95%

Dodavatel: Apollo Scientific

Synonyms: H-β-(2-Thienyl)-Ala-OH, 3-(2-Thienyl)-L-alanine, (S)-2-Amino-3-(thiophen-2-yl)propanoic acid

OR14732-5G
APOSOR14732-5GEA 4270 CZK
APOSOR14732-5G
3-(2-Thienyl)-L-alanine 95%
3-(2-Thienyl)-L-alanine
Vzorec: C₇H₉NO₂S
MW: 171,214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064335
CAS číslo: 22951-96-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR