Tisk…

3-Nitro-L-phenylalanine 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-2-Amino-3-(3-nitrophenyl)propanoic acid, H-Phe(3-NO₂)-OH

OR14707-1G OR14707-5G
APOSOR14707-1GEA 2150 CZK
APOSOR14707-1G APOSOR14707-5G
3-Nitro-L-phenylalanine 97%
3-Nitro-L-phenylalanine
Vzorec: C₉H₁₀N₂O₄
MW: 210,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860661
CAS číslo: 19883-74-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR