N-(4-Bromobutyl)phthalimide 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(4-Bromobutyl)isoindoline-1,3-dione
OR1468-100G
APOSOR1468-100GEA
2530
CZK
APOSOR1468-100G
N-(4-Bromobutyl)phthalimide 98%
N-(4-Bromobutyl)phthalimide
Vzorec:
C₁₂H₁₂BrNO₂ MW: 282,14 g/mol Bod tání: 76…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005905 CAS číslo: 5394-18-3 EINECS: 226-401-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...