Tisk…

Fmoc-L-allylglycine 97%

Dodavatel: Apollo Scientific

Synonyms: Fmoc-Gly(allyl)-OH

OR14672-5G
APOSOR14672-5GEA 2150 CZK
APOSOR14672-5G
Fmoc-L-allylglycine 97%
Fmoc-L-allylglycine
Vzorec: C₂₀H₁₉NO₄
MW: 337,38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311749
CAS číslo: 146549-21-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR