Tisk…

Fmoc-Dab(Boc)-OH 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (2S)-2,4-Diaminobutanoic acid, 4-BOC, 2-FMOC protected

OR14669-5G OR14669-10G
APOSOR14669-5GEA 787 CZK
APOSOR14669-5G APOSOR14669-10G
Fmoc-Dab(Boc)-OH 97%
Fmoc-Dab(Boc)-OH
Vzorec: C₂₄H₂₈N₂O₆
MW: 440,496 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151941
CAS číslo: 125238-99-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR