Tisk…

Fmoc-Dap(Boc)-OH 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Fmoc-(N-beta-boc)-L-alpha,beta-diaminopropionic acid, 3-(tert-Butoxycarbonyl)amino-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-alanine, (S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic acid, Nα-Fmoc-Nβ-Boc-L-2,3-diaminopropionic acid, (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 3-(Boc-amino)-N-Fmoc-L-alanine

OR14667-100G OR14667-25G
APOSOR14667-100GEA 16440 CZK
APOSOR14667-100G APOSOR14667-25G
Fmoc-Dap(Boc)-OH 97%
Fmoc-Dap(Boc)-OH
Vzorec: C₂₃H₂₆N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00235896
CAS číslo: 162558-25-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR