Tisk…

Fmoc-3-(3-pyridyl)-L-alanine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Fmoc-3-Pal-OH, Fmoc-β-(3-pyridyl)-Ala-OH

OR14660-25G OR14660-5G
APOSOR14660-25GEA 9860 CZK
APOSOR14660-25G APOSOR14660-5G
Fmoc-3-(3-pyridyl)-L-alanine 97%
Fmoc-3-(3-pyridyl)-L-alanine
Vzorec: C₂₃H₂₀N₂O₄
MW: 388,423 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144887
CAS číslo: 175453-07-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR