Tisk…
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid
Dodavatel: Apollo Scientific
Synonyms: N-Boc-3-(2-pyridyl)-L-alanine, Boc-2-Pal-OH
OR14654-1G OR14654-250MG
APOSOR14654-1GEA 910 CZK
APOSOR14654-1G APOSOR14654-250MG
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid
Vzorec: C₁₃H₁₈N₂O₄
MW: 266.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191190
CAS číslo: 71239-85-5