Tisk…

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine

Dodavatel: Apollo Scientific

Synonyms: Fmoc-beta-(2-furyl)-Ala-OH, Fmoc-3-(2-Furyl)-Ala-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid, N-Fmoc-3-(2-furyl)-L-alanine

OR14652-1G OR14652-5G
APOSOR14652-1GEA 2150 CZK
APOSOR14652-1G APOSOR14652-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine
Vzorec: C₂₂H₁₉NO₅
MW: 377,40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311747
CAS číslo: 159611-02-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR