Fmoc-4-amino-L-phenylalanine 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Fmoc-Phe(4-NH₂)-OH, 4-Amino-N-Fmoc-L-phenylalanine, Fmoc-Phe(4-NH2)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-aminophenyl)propanoic acid, Fmoc-4-amino-Phe-OH
OR14620-5G
OR14620-25G
APOSOR14620-5GEA
2610
CZK
APOSOR14620-5G
APOSOR14620-25G
Fmoc-4-amino-L-phenylalanine 95%
Fmoc-4-amino-L-phenylalanine
Vzorec:
C₂₄H₂₂N₂O₄ MW: 402,45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00237664 CAS číslo: 95753-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...