Tisk…

N-Fmoc-4-iodo-D-phenylalanine

Dodavatel: Apollo Scientific
Varování

OR14613-1G OR14613-5G
APOSOR14613-1GEA 1740 CZK
APOSOR14613-1G APOSOR14613-5G
N-Fmoc-4-iodo-D-phenylalanine
N-Fmoc-4-iodo-D-phenylalanine
Vzorec: C₂₄H₂₀INO₄
MW: 513,331 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672557
CAS číslo: 205526-29-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR