Tisk…

Fmoc-2,4-dichloro-L-phenylalanine

Dodavatel: Apollo Scientific

Synonyms: Fmoc-Phe(2,4-DiCl)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid, 2,4-Dichloro-L-phenylalanine, N-FMOC protected

OR14604-25G OR14604-5G
APOSOR14604-25GEA 10980 CZK
APOSOR14604-25G APOSOR14604-5G
Fmoc-2,4-dichloro-L-phenylalanine
Fmoc-2,4-dichloro-L-phenylalanine
Vzorec: C₂₄H₁₉Cl₂NO₄
MW: 456,33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863048
CAS číslo: 352351-62-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR