Fmoc-D-Tic-OH
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Fmoc-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
OR14590-25G
APOSOR14590-25GEA
2320
CZK
APOSOR14590-25G
Fmoc-D-Tic-OH
Fmoc-D-Tic-OH
Vzorec:
C₂₅H₂₁NO₄ MW: 399,446 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00144345 CAS číslo: 130309-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...