Tisk…

Boc-2-nitro-L-phenylalanine

Dodavatel: Apollo Scientific

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-(2-nitrophenyl)propanoic acid, Boc-Phe(2-NO₂)-OH, 2-Nitro-L-phenylalanine, N-BOC protected

OR14576-1G OR14576-5G
APOSOR14576-1GEA 2650 CZK
APOSOR14576-1G APOSOR14576-5G
Boc-2-nitro-L-phenylalanine
Boc-2-nitro-L-phenylalanine
Vzorec: C₁₄H₁₈N₂O₆
MW: 310,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317708
CAS číslo: 185146-84-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR