Tisk…

N-Boc-3-methyl-L-phenylalanine

Dodavatel: Apollo Scientific

Synonyms: Boc-Phe(3-Me)-OH

OR14568-5G
APOSOR14568-5GEA 3270 CZK
APOSOR14568-5G
N-Boc-3-methyl-L-phenylalanine
N-Boc-3-methyl-L-phenylalanine
Vzorec: C₁₅H₂₁NO₄
MW: 279,336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862944
CAS číslo: 114873-06-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR