Tisk…

4,5-Methylenedioxy-2-nitrocinnamic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(6-Nitro-1,3-benzodioxol-5-yl)acrylic acid

OR1456-25G OR1456-5G
APOSOR1456-25GEA 20900 CZK
APOSOR1456-25G APOSOR1456-5G
4,5-Methylenedioxy-2-nitrocinnamic acid
4,5-Methylenedioxy-2-nitrocinnamic acid
Vzorec: C₁₀H₇NO₆
MW: 237.17 g/mol
Bod tání: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00063250
CAS číslo: 6315-90-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR