Tisk…

2-Chloro-N-fmoc-L-phenylalanine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Fmoc-Phe(2-Cl)-OH

OR14542-25G OR14542-5G
APOSOR14542-25GEA 9660 CZK
APOSOR14542-25G APOSOR14542-5G
2-Chloro-N-fmoc-L-phenylalanine 97%
2-Chloro-N-fmoc-L-phenylalanine
Vzorec: C₂₄H₂₀ClNO₄
MW: 421,877 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00797578
CAS číslo: 198560-41-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR