Tisk…

(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone

Dodavatel: Apollo Scientific

Synonyms: (4S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone, (S)-4-Benzyl-3-propionyloxazolidin-2-one

OR14533-25G
APOSOR14533-25GEA 2150 CZK
APOSOR14533-25G
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone
Vzorec: C₁₃H₁₅NO₃
MW: 233,27 g/mol
Bod tání: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00269688
CAS číslo: 101711-78-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR