3-(Bromomethyl)benzaldehyde 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR1453-1G
OR1453-250MG
OR1453-5G
APOSOR1453-1GEA
2820
CZK
APOSOR1453-1G
APOSOR1453-250MG
APOSOR1453-5G
3-(Bromomethyl)benzaldehyde 97%
3-(Bromomethyl)benzaldehyde
Vzorec:
C₈H₇BrO Bod tání: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797497 CAS číslo: 82072-23-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...