2-Amino-3-nitrophenol
Dodavatel: Apollo Scientific
|
Varování
|
OR14485-25G
OR14485-5G
APOSOR14485-25GEA
744
CZK
APOSOR14485-25G
APOSOR14485-5G
2-Amino-3-nitrophenol
2-Amino-3-nitrophenol
Vzorec:
C₆H₆N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00010875 CAS číslo: 603-85-0 EINECS: 210-060-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...