Tisk…

4-(4-Chlorophenyl)-4-oxo-2-pyrrolidin-1-ylbutanoic acid

Dodavatel: Apollo Scientific

OR14472-1G OR14472-250MG
APOSOR14472-1GEA 0 CZK
APOSOR14472-1G APOSOR14472-250MG
4-(4-Chlorophenyl)-4-oxo-2-pyrrolidin-1-ylbutanoic acid
4-(4-Chlorophenyl)-4-oxo-2-pyrrolidin-1-ylbutanoic acid
Vzorec: C₁₄H₁₆ClNO₃
MW: 281,74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00954911
CAS číslo: 139084-68-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR