Tisk…

4,5-Dibenzyl-4H-1,2,4-triazole-3-thiol

Dodavatel: Apollo Scientific
Varování

OR14452-250MG OR14452-50MG
APOSOR14452-250MGEA 744 CZK
APOSOR14452-250MG APOSOR14452-50MG
4,5-Dibenzyl-4H-1,2,4-triazole-3-thiol
4,5-Dibenzyl-4H-1,2,4-triazole-3-thiol
Vzorec: C₁₆H₁₅N₃S
Storage Temperature: Ambient
MDL Number: MFCD00837917
CAS číslo: 83673-68-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR