Methyl-3-(4-hydroxyphenyl)acrylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 3-(4-hydroxyphenyl)acrylate, Methyl 4-hydroxycinnamate
OR14414-100G
OR14414-25G
OR14414-5G
APOSOR14414-100GEA
5100
CZK
APOSOR14414-100G
APOSOR14414-25G
APOSOR14414-5G
Methyl-3-(4-hydroxyphenyl)acrylate
Methyl-3-(4-hydroxyphenyl)acrylate
Vzorec:
C₁₀H₁₀O₃ MW: 178,1846 g/mol Bod tání: 137…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00157167 CAS číslo: 3943-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...