Tisk…

5-(4-Methylphenyl)-1H-tetrazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-(p-Tolyl)-1H-tetrazole

OR14403-1G OR14403-250MG
APOSOR14403-1GEA 1910 CZK
APOSOR14403-1G APOSOR14403-250MG
5-(4-Methylphenyl)-1H-tetrazole
5-(4-Methylphenyl)-1H-tetrazole
Vzorec: C₈H₈N₄
Storage Temperature: Ambient
MDL Number: MFCD01318164
CAS číslo: 24994-04-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR