Tisk…

3-Hydroxy-4'-methoxyflavone

Dodavatel: Apollo Scientific

Synonyms: 3-Hydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxyflavonol, 3-Hydroxy-2-(4-methoxyphenyl)-4-benzopyrone, 3-Hydroxy-2-(4-methoxyphenyl)chromen-4-one

OR14402-1G OR14402-25G OR14402-5G
APOSOR14402-1GEA 1240 CZK
APOSOR14402-1G APOSOR14402-25G APOSOR14402-5G
3-Hydroxy-4'-methoxyflavone
3-Hydroxy-4'-methoxyflavone
Vzorec: C₁₆H₁₂O₄
MW: 268,27 g/mol
Bod tání: 234 °C
Storage Temperature: Ambient
MDL Number: MFCD00017682
CAS číslo: 6889-78-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR