Tisk…

2-(1-Pyrrolyl)benzonitrile

Dodavatel: Apollo Scientific
Varování

OR1437-1G OR1437-5G
APOSOR1437-1GEA 1330 CZK
APOSOR1437-1G APOSOR1437-5G
2-(1-Pyrrolyl)benzonitrile
2-(1-Pyrrolyl)benzonitrile
Vzorec: C₁₁H₈N₂
Storage Temperature: Ambient
MDL Number: MFCD00128308
CAS číslo: 33265-71-3
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR