7-Bromoisatin 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione
OR14358-25G
OR14358-5G
OR14358-100G
APOSOR14358-25GEA
2820
CZK
APOSOR14358-25G
APOSOR14358-5G
APOSOR14358-100G
7-Bromoisatin 95%
7-Bromoisatin
Vzorec:
C₈H₄BrNO₂ MW: 226,03 g/mol Bod tání: 191…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00774354 CAS číslo: 20780-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...