tert-Butyl-4-(hydroxymethyl)benzylcarbamate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Aminomethyl)benzyl alcohol, N-BOC protected, 4-(Boc-aminomethyl-phenyl)-methanol, tert-Butyl 4-(hydroxymethyl)benzylcarbamate
OR14352-1G
OR14352-25G
OR14352-5G
APOSOR14352-1GEA
744
CZK
APOSOR14352-1G
APOSOR14352-25G
APOSOR14352-5G
tert-Butyl-4-(hydroxymethyl)benzylcarbamate 97%
tert-Butyl-4-(hydroxymethyl)benzylcarbamate
Vzorec:
C₁₃H₁₉NO₃ MW: 237,3 g/mol Bod tání: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD06411553 CAS číslo: 123986-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...