Dimethylglyoxim 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime
OR14325-100G
OR14325-2.5KG
OR14325-500G
APOSOR14325-100GEA
622
CZK
APOSOR14325-100G
APOSOR14325-2.5KG
APOSOR14325-500G
Dimethylglyoxim 99%
Dimethylglyoxim
Vzorec:
CH₃C(=NOH)C(=NOH)CH₃ MW: 116,12 g/mol Bod tání: 240…241 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002117 CAS číslo: 95-45-4 EINECS: 202-420-1 UN: 2926 ADR: 4.1+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...