Tisk…

4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid > 95%

Dodavatel: Apollo Scientific
Varování

OR14294-250MG
APOSOR14294-250MGEA 0 CZK
APOSOR14294-250MG
4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid > 95%
4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid
Vzorec: C₁₂H₁₃NO₃
MW: 219,24 g/mol
Bod tání: 169…171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06411619
CAS číslo: 2971-18-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR