1-(Methylsulphonyl)-4-piperidinecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(Methylsulphonyl)piperidine-4-carboxylic acid
OR14287-250MG
APOSOR14287-250MGEA
1660
CZK
APOSOR14287-250MG
1-(Methylsulphonyl)-4-piperidinecarboxylic acid
1-(Methylsulphonyl)-4-piperidinecarboxylic acid
Vzorec:
C₇H₁₃NO₄S MW: 207,2500 g/mol Bod tání: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD03821886 CAS číslo: 280772-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...