Tisk…

Tetrahydro-2H-pyran-4-carbonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR14254-5G OR14254-10G OR14254-1G
APOSOR14254-5GEA 1910 CZK
APOSOR14254-5G APOSOR14254-10G APOSOR14254-1G
Tetrahydro-2H-pyran-4-carbonyl chloride 97%
Tetrahydro-2H-pyran-4-carbonyl chloride
Vzorec: C₆H₉ClO₂
MW: 148,5875 g/mol
Bod varu: 50…52 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD06200863
CAS číslo: 40191-32-0
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR