Tisk…

4-(1-Pyrrolidinyl)benzenecarboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(1-Pyrrolidinyl)benzoic acid, 4-(Pyrrolidin-1-yl)benzoic acid

OR14246-250MG OR14246-5G
APOSOR14246-250MGEA 622 CZK
APOSOR14246-250MG APOSOR14246-5G
4-(1-Pyrrolidinyl)benzenecarboxylic acid
4-(1-Pyrrolidinyl)benzenecarboxylic acid
Vzorec: C₁₁H₁₃NO₂
MW: 191,23 g/mol
Bod tání: 262 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631241
CAS číslo: 22090-27-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR