Tisk…

Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl-2-chloro-2-2-4-methylphenyl-hydrazono-acetate, Ethyl-2-chloro-2-[2-(4-methylphenyl)hydrazono]acetate, Ethyl chloro[(4-methylphenyl)hydrazono]acetate

OR14210-1G
APOSOR14210-1GEA 3940 CZK
APOSOR14210-1G
Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate
Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate
Vzorec: C₁₁H₁₃ClN₂O₂
MW: 240,69 g/mol
Bod tání: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00266044
CAS číslo: 27171-88-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR