5-(4-Benzylpiperazino)-2-nitroaniline
Dodavatel: Apollo Scientific
OR14189-1G
OR14189-5G
APOSOR14189-1GEA
1660
CZK
APOSOR14189-1G
APOSOR14189-5G
5-(4-Benzylpiperazino)-2-nitroaniline
5-(4-Benzylpiperazino)-2-nitroaniline
Vzorec:
C₁₇H₂₀N₄O₂ Bod tání: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200970 CAS číslo: 23470-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...