Tisk…

4-Formylbenzeneboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Formylphenylboronic acid pinacol ester, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

OR14173-100G OR14173-25G OR14173-5G
APOSOR14173-100GEA 19350 CZK
APOSOR14173-100G APOSOR14173-25G APOSOR14173-5G
4-Formylbenzeneboronic acid pinacol ester 98%
4-Formylbenzeneboronic acid pinacol ester
Vzorec: C₁₃H₁₇BO₃
MW: 232.086 g/mol
Bod tání: 54…59 °C
Storage Temperature: Ambient
MDL Number: MFCD04972375
CAS číslo: 128376-64-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR