Isobutylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methylpropylboronic acid, (2-Methylpropyl)boronic acid
OR14160-100G
OR14160-25G
OR14160-5G
APOSOR14160-100GEA
2860
CZK
APOSOR14160-100G
APOSOR14160-25G
APOSOR14160-5G
Isobutylboronic acid
Isobutylboronic acid
Vzorec:
C₄H₁₁BO₂ MW: 101.94 Bod tání: 108…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134156 CAS číslo: 84110-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...