Tisk…

4-Isoquinolineboronic acid 96%

Dodavatel: Apollo Scientific
Varování

OR14158-1G OR14158-5G
APOSOR14158-1GEA 2860 CZK
APOSOR14158-1G APOSOR14158-5G
4-Isoquinolineboronic acid 96%
4-Isoquinolineboronic acid
Vzorec: C₉H₈BNO₂
MW: 172.9763 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03412093
CAS číslo: 192182-56-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR